Learn More
R&D Systems™ Human TNF-alpha Quantikine QuicKit Control Group 259
Shop All Novus Biologicals Products

Description
- For use as quantitative controls for the determination of human TNF-α concentrations in biological fluids.
- Concentrations have been assigned using the R&D Systems™ Human TNF-α Quantikine™ QuicKit™ ELISA kit.
- Controls are prepared in a buffered protein base with preservatives.
- They contain recombinant human TNF-α at low and high concentrations.
- Controls are supplied lyophilized.
Specifications
Specifications
| Product Type | ELISA |
| For Use With (Equipment) | Catalog number QK210 |
| Gene ID (Entrez) | 7124 |
| Host Species | Human |
| Quantity | 1 Each |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
| Target | TNF-alpha |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.