Learn More
Description
- A one step, 80 minute solid phase ELISA designed to measure human TNF-alpha in cell culture supernates, serum, and plasma.
- It contains E. coli-expressed recombinant human TNF-alpha and antibodies raised against the recombinant protein.
- Results obtained for naturally occurring human TNF-alpha showed linear curves that were parallel to the standard curves obtained using the recombinant QuicKit™ standards.
- These results indicate that this kit can be used to determine relative mass values for natural human TNF-alpha.
Specifications
Specifications
| Product Type | ELISA |
| Specificity | Natural and Recombinant Human TNF-alpha |
| Gene ID (Entrez) | 7124 |
| Host Species | Human |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
| Target | TNF-alpha |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
