Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TNF-beta Recombinant Protein, PeproTech®

Catalog No. 30001B01M
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-001-B01M 2 x 500 μg
30001B100UG 100 μg
30001B20UG 20 μg
30001B250UG 250 μg
30001B500UG 500 μg
30001B50UG 50 μg
300-01B-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 30-001-B01M Supplier Gibco™ Supplier No. 30001B01M
Add to Cart
Add to Cart

Recombinant Protein

300-01B-1MG will be provided as 2 x 500 μg (300-01B-500UG). Recombinant Human TNF-beta is a secreted 172 amino acid protein (18.6 kDa) which forms an active non-disulfide-linked homotrimer structure in solution. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TNF beta, a member of the tumor necrosis factor family, is a cytokine produced by lymphocytes. The protein is highly inducible, secreted, and forms heterotrimers with lymphotoxin-beta which anchor lymphotoxin-alpha to the cell surface. This protein also mediates a large variety of inflammatory, immunostimulatory, and antiviral responses, is involved in the formation of secondary lymphoid organs during development and plays a role in apoptosis. Genetic variations in this gene are associated with susceptibility to leprosy type 4 and psoriatic arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01374
For Use With (Application) Control, Western Blot Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 4049
Molecular Weight (g/mol) 18.6 kDa
Name Human TNF-beta
Purification Method Purified
Quantity 2 x 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias DAMA-25N12.13; hlb382; hypothetical protein; LT; LT[a]; LT-[a]; LTA; LTalpha; LT-alpha; Ltx; lymphotoxin A; lymphotoxin alpha; lymphotoxin alpha (TNF superfamily, member 1); lymphotoxin-alpha; lymphotoxin-alpha; lymphotoxin alpha; TNF b; TNF beta; TNF superfamily, member 1; TNF β; Tnfb; TNFbeta; TNF-beta; TNF-N; Tnfsf1; Tnfsf1b; TNFβ; TNLG1E; tumor necrosis factor beta; tumor necrosis factor ligand 1E; tumor necrosis factor ligand superfamily member 1
Common Name TNF beta
Gene Symbol LTA
Biological Activity The ED50 as determined by the cytolysis of murine L929 cells in the presence of Actinomycin-D is ≤ 0.05 ng/ml, corresponding to a specific activity of ≥ 2 x 10^7 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MLPGVGLTPS AAQTARQHPK MHLAHSTLKP AAHLIGDPSK QNSLLWRANT DRAFLQDGFS LSNNSLLVPT SGIYFVYSQV VFSGKAYSPK ATSSPLYLAH EVQLFSSQYP FHVPLLSSQK MVYPGLQEPW LHSMYHGAAF QLTQGDQLST HTDGIPHLVL SPSTVFFGAF AL
Content And Storage -20°C
Activity The ED50 as determined by the cytolysis of murine L929 cells in the presence of Actinomycin-D is ≤ 0.05 ng/ml, corresponding to a specific activity of ≥ 2 x 10^7 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.