Learn More
Kingfisher Biotech Inc Human TNF-R1 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1807H005
Encompass
The Human TNF-R1 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Human TNF-R1 applications are for cell culture, ELISA standard, and Western Blot Control. Human TNF-R1 yeast-derived recombinant protein can be purchased in multiple sizes. Human TNF-R1 Specifications: (Molecular Weight: 20.5 kDa) (Amino Acid Sequence: LVPHLGDREK RDSVCPQGKY IHPQNNSICC TKCHKGTYLY NDCPGPGQDT DCRECESGSF TASENHLRHC LSCSKCRKEM GQVEISSCTV DRDTVCGCRK NQYRHYWSEN LFQCFNCSLC LNGTVHLSCQ EKQNTVCTCH AGFFLRENEC VSCSNCKKSL ECTKLCLPQI ENVKGTEDSG TT (182)) (Gene ID: 7132). For research use only. Made in the USA
Catalog No. 50-253-2740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.