Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human TNF RI/TNFRSF1A Quantikine QuicKit Control Group 270
Shop All Novus Biologicals Products

Specifications
Specifications
| For Use With (Application) | Control |
| Gene ID (Entrez) | 7132 |
| Host Species | Human |
| Quantity | 1 Ea. |
| Storage Requirements | Store the unopened product at 2 to 8 °C. Do not use past expiration date |
| Target | TNF RI/TNFRSF1A |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?