Learn More
Specifications
Specifications
| Assay Range | 15.60 to 1000 pg/mL |
| Assay Sensitivity | 1.69 pg/mL |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Heparin Plasma, Serum, Urine |
| Specificity | Natural and Recombinant Human TNF RI. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7132 |
| Host Species | Human |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 to 8 °C. Do not use past expiration date |
| Synonym | CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNFRI, TNF-RI, TNF-R-I, TNFR-I, TNFRSF1A, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1 A isoform beta, tumor necrosis factor receptor superfamily member 1 A, tumor necrosis factor receptor superfamily, member 1 A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
