Learn More
Sartorius Human TNFa T Cell Companion
Supplier:  Sartorius 97034
Encompass
The iQue Human T Cell Companion Kits can be used to measure additional cytokines besides those already included in the Human T Cell Immunology portfolio of Kits.
Catalog No. 50-289-1731