Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TNFAIP2 Synthetic Peptide

Catalog No. PEP1498
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1498 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1498 Supplier Invitrogen™ Supplier No. PEP1498

Synthetic Peptide

TNFAIP2 Synthetic Peptide, PEP-1498, from Invitrogen.

TNFAIP2, also known as B94, is a TNF-a-regulated gene that is expressed in endothelial cells, peripheral blood cells, and mature sperm. Recently, elevated TNFAIP2 expression was observed in nasopharyngeal carcinoma, and high expression of TNFAIP2 was significantly correlated with higher levels of cell migration, invasion and metastasis, suggesting that TNFAIP2 may serve as a useful prognostic indicator for nasopharyngeal carcinoma. TNFAIP2 has also been implicated as part of the viral-sensing circuit of the innate immune response.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 7127
Species Human
Purification Method purified
Accession Number Q03169
Common Name Human TNFAIP2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol TNFAIP2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.