Learn More
Sino Biological Human TNFRSF10C MAb 25TESTS 10415-R003-A-25
Supplier:  Sino Biological 10415R003A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TRAILR3/TNFRSF10C (rh TRAILR3/TNFRSF10C; Catalog#10415-H08H; NP_003832.2; Met1-Pro235) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-0607
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.