Learn More
Sino Biological Human TNFRSF1A PAb 100UL 10872-T16-100
Supplier:  Sino Biological 10872T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TNFR1 / CD120a / TNFRSF1A (rh TNFR1 / CD120a / TNFRSF1A; Catalog#10872-H08H; NP_001056.1; Met1-Thr211). TNFR1 / CD120a / TNFRSF1A specific IgG was purified by Human TNFR1 / CD120a / TNFRSF1A affinity chromatography.
Catalog No. 50-240-9414
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.