Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TNFRSF25 Recombinant Protein fused to murine IgG2a Fc and hinge region purifird from CHO cells

Catalog No. 89965230 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-230 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-965-230 Supplier Abnova™ Supplier No. P4936
Add to Cart
Add to Cart

Used for SDS-PAGE

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is expressed preferentially in the tissues enriched in lymphocytes, and it may play a role in regulating lymphocyte homeostasis. This receptor has been shown to stimulate NF-kappa B activity and regulate cell apoptosis. The signal transduction of this receptor is mediated by various death domain containing adaptor proteins. Knockout studies in mice suggested the role of this gene in the removal of self-reactive T cells in the thymus. Multiple alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported, most of which are potentially secreted molecules. The alternative splicing of this gene in B and T cells encounters a programmed change upon T-cell activation, which predominantly produces full-length, membrane bound isoforms, and is thought to be involved in controlling lymphocyte proliferation induced by T-cell activation. [provided by RefSeq]

Sequence: N-teminal Sequence: GTRSP

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 8718
Molecular Weight (g/mol) 42.9kDa
Name TNFRSF25 (Human) Recombinanat Protein
Preparation Method Mammalian cell (CHO) expression system
Purification Method Affinity and size purification
Quantity 25 μg
Immunogen N-teminal sequence: GTRSP
Storage Requirements Store at 4°C. Open under aseptic conditions. This product is stable for at least 3 months.
Regulatory Status RUO
Gene Alias APO-3/DDR3/DR3/LARD/TNFRSF12/TR3/TRAMP/WSL-1/WSL-LR
Common Name TNFRSF25
Gene Symbol TNFRSF25
Species Human
Recombinant Recombinant
Protein Tag None
Expression System Mammalian cell (CHO) expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.