Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TNFRSF4 Recombinant Protein fused to murine IgG2a Fc purifird from CHO cells

Catalog No. 89965233 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-233 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-965-233 Supplier Abnova™ Supplier No. P4934
Add to Cart
Add to Cart

Used for SDS-PAGE

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor has been shown to activate NF-kappaB through its interaction with adaptor proteins TRAF2 and TRAF5. Knockout studies in mice suggested that this receptor promotes the expression of apoptosis inhibitors BCL2 and BCL2lL1/BCL2-XL, and thus suppresses apoptosis. The knockout studies also suggested the roles of this receptor in CD4+ T cell response, as well as in T cell-dependent B cell proliferation and differentiation. [provided by RefSeq]

Sequence: TNFRSF4 mature: lhcvgdtypsndrcchecrpgngmvsrcsrsqntvcrpcgpgfyndvvsskpckpctwcnlrsgserkqlctatqdtvcrcragtqpldsykpgvdcapcppghfspgdnqackpwtnctlagkhtlqpasnssdaicedrdppatqpqetqgpparpitvqpteawprtsqgpstr

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 7293
Molecular Weight (g/mol) 45.6kDa
Name TNFRSF4 (Human) Recombinanat Protein
Preparation Method Mammalian cell (CHO) expression system
Purification Method Size purification
Quantity 25 μg
Immunogen TNFRSF4 mature: lhcvgdtypsndrcchecrpgngmvsrcsrsqntvcrpcgpgfyndvvsskpckpctwcnlrsgserkqlctatqdtvcrcragtqpldsykpgvdcapcppghfspgdnqackpwtnctlagkhtlqpasnssdaicedrdppatqpqetqgpparpitvqpteawprtsqgpstr Fused to murine IgG2a Fc: eprgptikpcppckcpapnllggpsvfifppkikdvlmislspivtcvvvdvseddpdvqiswfvnnvevhtaqtqthredynstlrvvsalpiqhqdwmsgkefkckvnnkdlpapiertiskpkgsvrapqvyvlpppeeemtkkqvtltcmvtdfmpediyvewtnngktelnykntepvldsdgsyfmysklrvekknwvernsyscsvvheglhnhhttksfsrtpg
Storage Requirements Store at 4°C. This product is stable for at least 3 months.
Regulatory Status RUO
Gene Alias ACT35/CD134/OX40/TXGP1L
Common Name TNFRSF4
Gene Symbol TNFRSF4
Species Human
Recombinant Recombinant
Protein Tag None
Expression System Mammalian cell (CHO) expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.