Learn More
Proteintech Group Inc Human TNFRSF9 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0012596T
Encompass_Preferred
CD137, also known as TNFRSF9 or 4-1BB, is an inducible T cell surface receptor which belongs to the tumor necrosis factor receptor superfamily. CD137 is a transmembrane protein expressed on the surface of activated T-cells. In addition, activation-dependent expression of CD137 has also been found in B lymphocytes, monocytes, and diverse nonlymphoid cell types. CD137 provides a co-stimulatory signal that enhances the survival, and differentiation of cells, and has a crucial role in the development of CD8 cytotoxic T cells and anti-tumor immunity. Soluble forms of CD137 (sCD137) are generated by differential splicing and are released by activated lymphocytes. Elevated serum sCD137 level has been associated with some diseases, including rheumatoid arthritis, multiple sclerosis, chronic lymphocytic leukemia, and acute coronary syndrome.
Catalog No. 50-197-4084
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.