Learn More
ACROBiosystems Human TNFSF11 / RANKL / CD254 Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems RALH52A650UG
Encompass_Preferred
Recombinant Protein;50UG;Human TNFSF11, Mouse IgG2a Fc Tag (RAL-H52A6) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Accession # AAC51762.1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 47.4 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Osteoprotegerin, His Tag (Cat. No. TNB-H5220) at 2 μg/mL (100 μL/well) can bind Human TNFSF11, Mouse IgG2a Fc Tag (Cat. No. RAL-H52A6) with a linear range of 0.2-2 ng/mL (QC tested).;Immobilized Human TNFSF11, Mouse IgG2a Fc Tag (Cat. No. RAL-H52A6) at 2 μg/mL (100 μL/well) can bind Anti-Human TNFSF11 Antibody, Human IgG1 with a linear range of 0.2-2 ng/mL (Routinely tested).
Catalog No. 50-205-5032
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.