Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TNS1 Full-length ORF (AAH51304.1, 1 a.a. - 385 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965375 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-375 25 ug
89-965-374 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-375 Supplier Abnova™ Supplier No. H00007145P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene localizes to focal adhesions, regions of the plasma membrane where the cell attaches to the extracellular matrix. This protein crosslinks actin filaments and contains a Src homology 2 (SH2) domain, which is often found in molecules involved in signal transduction. This protein is a substrate of calpain II. A second transcript from this gene has been described, but its full length nature has not been determined. [provided by RefSeq]

Sequence: MSVSRTMEDSCELDLVYVTERIIAVSFPSTANEENFRSNLREVAQMLKSKHGGNYLLFNLSERRPDITKLHAKVLEFGWPDLHTPALEKICSICKAMDTWLNADPHNVVVLHNKGNRGRIGVVIAAYMHYSNISASADQALDRFAMKRFYEDKIVPIGQPSQRRYVHYFSGLLSGSIKMNNKPLFLHHVIMHGIPNFESKGGCRPFLRIYQAMQPVYTSGIYNIPGDSQTSVCITIEPGLLLKGDILLKCYHKKFRSPARDVIFRVQFHTCAIHDLGVVFGKEDLDDAFKDDRFPEYGKVEFVFSYGPEKIQGMEHLENGPSVSVDYNTSDPLIRWDSYDNFSGHRDDGMEDGNKQNTNSQSIGSISGGLEDQYTWPDTHWPSQS

Specifications

Accession Number AAH51304.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7145
Molecular Weight (g/mol) 70.1kDa
Name TNS1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MSVSRTMEDSCELDLVYVTERIIAVSFPSTANEENFRSNLREVAQMLKSKHGGNYLLFNLSERRPDITKLHAKVLEFGWPDLHTPALEKICSICKAMDTWLNADPHNVVVLHNKGNRGRIGVVIAAYMHYSNISASADQALDRFAMKRFYEDKIVPIGQPSQRRYVHYFSGLLSGSIKMNNKPLFLHHVIMHGIPNFESKGGCRPFLRIYQAMQPVYTSGIYNIPGDSQTSVCITIEPGLLLKGDILLKCYHKKFRSPARDVIFRVQFHTCAIHDLGVVFGKEDLDDAFKDDRFPEYGKVEFVFSYGPEKIQGMEHLENGPSVSVDYNTSDPLIRWDSYDNFSGHRDDGMEDGNKQNTNSQSIGSISGGLEDQYTWPDTHWPSQS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp586K0617/MGC88584/MST091/MST122/MST127/MSTP122/MSTP127/MXRA6/TNS
Common Name TNS1
Gene Symbol TNS1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.