Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Total ErbB3/Her3 DuoSet IC ELISA
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
Description
The Human Total ErbB3/Her3 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human ErbB3/Her3.
Specifications
Specifications
Product Type | ELISA Development Kit for Intracellular Proteins |
Quantity | 1 Kit |
Species | Human |
Conjugate | Biotin |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 2065 |
Packaging Quantity | 2 Plates |
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction