Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total IL-18/IL-1F4 Quantikine ELISA Kit
SDP

Catalog No. DL180 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DL180 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DL180 Supplier R&D Systems™ Supplier No. DL180
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Total IL-18/IL-1F4 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Total IL-18/IL-1F4 Quantikine ELISA Kit quantifies human IL-18/IL-1F4 in cell culture supernates (50 ul), serum (17 ul), edta plasma (17 ul), heparin plasma (17 ul), saliva (25 ul), urine (50 ul).

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 5.15pg/mL
Product Line R&D Systems
Product Type Total IL-18/IL-1F4 Quantikine ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human Total IL-18
For Use With (Application) ELISA
Gene ID (Entrez) 3606
Host Species Human
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (17uL),EDTA Plasma (17uL),Heparin Plasma (17uL),Saliva (25uL),Urine (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18 mgC12320, IL-1F4, IL1F4iboctadekin, IL-1 g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Target IL-18/IL-1F4
Test Time 4 hr.
Show More Show Less

For Research Use Only. Not for use in Humans.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.