Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total MMP-1 DuoSet ELISA
SDP

Catalog No. DY901B05 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY901B05 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY901B05 Supplier R&D Systems™ Supplier No. DY901B05
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Total MMP-1 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Total MMP-1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human MMP-1.

Specifications

Assay Range 62.50 to 4000 pg/mL
Product Type Solid Phase Sandwich ELISA
For Use With (Application) ELISA
Gene ID (Entrez) 4312
Host Species Human
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP1, MMP-1
Target MMP-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.