Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Human Total MMP-7 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human MMP-7 in cell culture supernates (25 ul), serum (25 ul), heparin plasma (25 ul), saliva (25 ul), urine (50 ul).
Specifications
Specifications
| Assay Range | 0.16 to 10 ng/mL |
| Assay Sensitivity | 0.094 ng/mL |
| Conjugate | HRP |
| Cross Reactivity | Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules. |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (Heparin), Saliva, Serum, Urine |
| Specificity | Natural and Recombinant Human MMP-7 |
| Sufficient For | 6 x 96-well Plates |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
