Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total MMP-8 Quantikine ELISA Kit
SDP

Catalog No. DMP800B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DMP800B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DMP800B Supplier R&D Systems™ Supplier No. DMP800B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Total MMP-8 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Total MMP-8 Quantikine ELISA Kit quantifies human MMP-8 in cell culture supernates (50 μL), tissue lysates (50 μL), serum (10 μL), edta plasma (25 μL), platelet-poor edta plasma (50 μL), heparin plasma (25 μL), platelet-poor heparin plasma (50 μL), saliva (10 μL).

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.058 ng/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Platelet-poor Plasma (EDTA, Heparin), Saliva, Serum, Tissue Lysates
Specificity Natural and Recombinant Human MMP-8
For Use With (Application) ELISA
Gene ID (Entrez) 4317
Host Species Human
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50 μL),Tissue Lysates (50 μL),Serum (10 μL),EDTA Plasma (25 μL),Platelet-poor EDTA Plasma (50 μL),Heparin Plasma (25 μL),Platelet-poor Heparin Plasma (50 μL),Saliva (10 μL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CLG1HNC, Collagenase 2, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL
Target MMP-8
Total Assay Time 4.5 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.