Learn More
Abnova™ Human TPSD1 Full-length ORF (NP_036349.1, 1 a.a. - 242 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
Tryptases comprise a family of trypsin-like serine proteases, the peptidase family S1. Tryptases are enzymatically active only as heparin-stabilized tetramers, and they are resistant to all known endogenous proteinase inhibitors. Several tryptase genes are clustered on chromosome 16p13.3. These genes are characterized by several distinct features. They have a highly conserved 3' UTR and contain tandem repeat sequences at the 5' flank and 3' UTR which are thought to play a role in regulation of the mRNA stability. Although this gene may be an exception, most of the tryptase genes have an intron immediately upstream of the initiator Met codon, which separates the site of transcription initiation from protein coding sequence. This feature is characteristic of tryptases but is unusual in other genes. Tryptases have been implicated as mediators in the pathogenesis of asthma and other allergic and inflammatory disorders. This gene was once considered to be a pseudogene, although it is now believed to be a functional gene that encodes a protein. [provided by RefSeq]
Specifications
Specifications
| Accession Number | NP_036349.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 23430 |
| Molecular Weight (g/mol) | 53kDa |
| Name | TPSD1 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 10 μg |
| Immunogen | MLLLAPQMLSLLLLALPVLASPAYVAPAPGQALQQTGIVGGQEAPRSKWPWQVSLRVRGPYWMHFCGGSLIHPQWVLTAAHCVEPDIKDLAALRVQLREQHLYYQDQLLPVSRIIVHPQFYIIQTGADIALLELEEPVNISSHIHTVTLPPASETFPPGMPCWVTGWGDVDNNVHLPPPYPLKEVEVPVVENHLCNAEYHTGLHTGHSFQIVRDDMLCAGSENHDSCQGDSGGPLVCKVNGT |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.