Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TPSD1 Full-length ORF (NP_036349.1, 1 a.a. - 242 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965510 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-510 25 μg
89-965-509 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-510 Supplier Abnova™ Supplier No. H00023430P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Tryptases comprise a family of trypsin-like serine proteases, the peptidase family S1. Tryptases are enzymatically active only as heparin-stabilized tetramers, and they are resistant to all known endogenous proteinase inhibitors. Several tryptase genes are clustered on chromosome 16p13.3. These genes are characterized by several distinct features. They have a highly conserved 3' UTR and contain tandem repeat sequences at the 5' flank and 3' UTR which are thought to play a role in regulation of the mRNA stability. Although this gene may be an exception, most of the tryptase genes have an intron immediately upstream of the initiator Met codon, which separates the site of transcription initiation from protein coding sequence. This feature is characteristic of tryptases but is unusual in other genes. Tryptases have been implicated as mediators in the pathogenesis of asthma and other allergic and inflammatory disorders. This gene was once considered to be a pseudogene, although it is now believed to be a functional gene that encodes a protein. [provided by RefSeq]

Sequence: MLLLAPQMLSLLLLALPVLASPAYVAPAPGQALQQTGIVGGQEAPRSKWPWQVSLRVRGPYWMHFCGGSLIHPQWVLTAAHCVEPDIKDLAALRVQLREQHLYYQDQLLPVSRIIVHPQFYIIQTGADIALLELEEPVNISSHIHTVTLPPASETFPPGMPCWVTGWGDVDNNVHLPPPYPLKEVEVPVVENHLCNAEYHTGLHTGHSFQIVRDDMLCAGSENHDSCQGDSGGPLVCKVNGT

Specifications

Accession Number NP_036349.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23430
Molecular Weight (g/mol) 53kDa
Name TPSD1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MLLLAPQMLSLLLLALPVLASPAYVAPAPGQALQQTGIVGGQEAPRSKWPWQVSLRVRGPYWMHFCGGSLIHPQWVLTAAHCVEPDIKDLAALRVQLREQHLYYQDQLLPVSRIIVHPQFYIIQTGADIALLELEEPVNISSHIHTVTLPPASETFPPGMPCWVTGWGDVDNNVHLPPPYPLKEVEVPVVENHLCNAEYHTGLHTGHSFQIVRDDMLCAGSENHDSCQGDSGGPLVCKVNGT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MCP7-LIKE/MCP7L1/MGC95428/MMCP-7L
Common Name TPSD1
Gene Symbol TPSD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.