Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRA16 Partial ORF (NP_795361, 40 a.a. - 139 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965533 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-533 25 μg
89-965-532 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-533 Supplier Abnova™ Supplier No. H00126382Q01L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for AP, Array, ELISA, WB-Re

Sequence: NSDQGPSQWVTLEFPQLIRVSQLQIQFQGGFSSRRGCLEGSQGTQALHKIVDFYPEDNNSLQTFPIPAAEVDRLKVTFEDATDFFGRVVIYHLRVLGEKV

Specifications

Accession Number NP_795361
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 126382
Molecular Weight (g/mol) 36.74kDa
Name TRA16 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen NSDQGPSQWVTLEFPQLIRVSQLQIQFQGGFSSRRGCLEGSQGTQALHKIVDFYPEDNNSLQTFPIPAAEVDRLKVTFEDATDFFGRVVIYHLRVLGEKV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias TRA16
Common Name NR2C2AP
Gene Symbol NR2C2AP
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.