Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRAF3IP1 Partial ORF (NP_056465.2, 592 a.a. - 690 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965554 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-554 10 μg
89-965-555 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-554 Supplier Abnova™ Supplier No. H00026146Q01S

Used for AP, Array, ELISA, WB-Re

Sequence: TLCKSALPLGKIMDYIQEDVDAMQNELQMWHSENRQHAEALQQEQRITDCAVEPLKAELAELEQLIKDQQDKICAVKANILKNEEKIQKMVYSINLTSR

Specifications

Accession Number NP_056465.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 26146
Molecular Weight (g/mol) 36.63kDa
Name TRAF3IP1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen TLCKSALPLGKIMDYIQEDVDAMQNELQMWHSENRQHAEALQQEQRITDCAVEPLKAELAELEQLIKDQQDKICAVKANILKNEEKIQKMVYSINLTSR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp434F124/MIP-T3/MIPT3
Common Name TRAF3IP1
Gene Symbol TRAF3IP1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.