Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRAF4 Partial ORF (AAH01769, 371 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965560 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-560 25 μg
89-965-559 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-560 Supplier Abnova™ Supplier No. H00009618Q01L

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the TNF receptor associated factor (TRAF) family. TRAF proteins are associated with, and mediate signal transduction from members of the TNF receptor superfamily. The encoded protein has been shown to interact with neurotrophin receptor, p75 (NTR/NTSR1), and negatively regulate NTR induced cell death and NF-kappa B activation. This protein has been found to bind to p47phox, a cytosolic regulatory factor included in a multi-protein complex known as NAD(P)H oxidase. This protein thus, is thought to be involved in the oxidative activation of MAPK8/JNK. Alternatively spliced transcript variants have been observed but the full-length nature of only one has been determined. [provided by RefSeq]

Sequence: PGAFDNLLEWPFARRVTFSLLDQSDPGLAKPQHVTETFHPDPNWKNFQKPGTWRGSLDESSLGFGYPKFISHQDIRKRNYVRDDAVFIRAAVELPRKILS

Specifications

Accession Number AAH01769
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9618
Molecular Weight (g/mol) 36.63kDa
Name TRAF4 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen PGAFDNLLEWPFARRVTFSLLDQSDPGLAKPQHVTETFHPDPNWKNFQKPGTWRGSLDESSLGFGYPKFISHQDIRKRNYVRDDAVFIRAAVELPRKILS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CART1/MLN62/RNF83
Common Name TRAF4
Gene Symbol TRAF4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.