Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRAF7 Full-length ORF (AAH24267.1, 1 a.a. - 594 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965572 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-572 10 μg
89-965-573 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-572 Supplier Abnova™ Supplier No. H00084231P01S

Used for AP, Array, ELISA, WB-Re

Tumor necrosis factor (TNF; see MIM 191160) receptor-associated factors, such as TRAF7, are signal transducers for members of the TNF receptor superfamily (see MIM 191190). TRAFs are composed of an N-terminal cysteine/histidine-rich region containing zinc RING and/or zinc finger motifs; a coiled-coil (leucine zipper) motif; and a homologous region that defines the TRAF family, the TRAF domain, which is involved in self-association and receptor binding.[supplied by OMIM]

Sequence: MPPISTPRRSDSAISVRSLHSESSMSLRSTFSLPEEEEEPEPLVFAEQPSVKLCCQLCCSVFKDPVITTCGHTFCRRCALKSEKCPVDNVKLTVVVNNIAVAEQIGELFIHCRHGCRVAGSGKPPIFEVDPRGCPFTIKLSARKDHEGSCDYRPVRCPNNPSCPPLLRMNLEAHLKECEHIKCPHSKYGCTFIGNQDTYETHLETCRFEGLKEFLQQTDDRFHEMHVALAQKDQEIAFLRSMLGKLSEKIDQLEKSLELKFDVLDENQSKLSEDLMEFRRDASMLNDELSHINARLNMGILGSYDPQQIFKCKGTFVGHQGPVWCLCVYSMGDLLFSGSSDKTIKVWDTCTTYKCQKTLEGHDGIVLALCIQGCKLYSGSADCTIIVWDIQNLQKVNTIRAHDNPVCTLVSSHNVLFSGSLKAIKVWDIVGTELKLKKELTGLNHWVRALVAAQSYLYSGSYQTIKIWDIRTLDCIHVLQTSGGSVYSIAVTNHHIVCGTYENLIHVWDIESKEQVRTLTGHVGTVYALAVISTPDQTKVFSASYDRSLRVWSMDNMICTQTLLRHQGSVTALAVSRGRLFSGAVDSTVKVWTC

Specifications

Accession Number AAH24267.1
For Use With (Application) Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84231
Molecular Weight (g/mol) 92.9kDa
Name Human TRAF7 Full-length ORF (AAH24267.1, 1 a.a. - 594 a.a.) Recombinant Protein with GST-tag at N-terminal
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Storage Requirements Store at −80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp586I021, MGC7807, RFWD1, RNF119
Common Name TRAF7
Gene Symbol TRAF7
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST Tag
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.