Learn More
Description
Specifications
Specifications
| Detectable Analytes | TRAIL/TNFSF10 |
| Assay | ELISA Development Kit |
| Conjugate | Biotin |
| Cross Reactivity | < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules. |
| Immunoassay Kit Format | 96-well microplate (sold separately) |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 8743 |
| Host Species | Human |
| Measuring Range | 23.4-1,500pg/mL |
| Quantity | 5 Plates |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
