Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TRANCE (RANKL) Recombinant Protein

Catalog No. PIRP8630
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP8630 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP8630 Supplier Invitrogen™ Supplier No. RP8630

Recombinant Protein

soluble RANK-L (human) Recombinant Protein, RP-8630, from Invitrogen.

Receptor Activator of NF-kB Ligand (RANKL) is a cell-bound marker (CD254) related to the TNF family of proteins. RANKL plays a critical role in bone metabolism, particularly osteoclast differentiation. In addition, RANKL is expressed by some T cells is thought to promote dendritic cell maturation. Recombinant soluble human RANKL is a non-glycosylated protein, containing 176 amino acids, with a molecular weight of 20 kDa. Purity is typically greater than 98% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the dose-dependent stimulation of IL-8 production in human PBMC and is typically less than 100 ng/mL. The specific activity of this protein is 1x10^4 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: MEKAMVDGSW LDLAKRSKLE AQPFAHLTIN ATDIPSGSHK VSLSSWYHDR GWAKISNMTF SNGKLIVNQD GFYYLYANIC FRHHETSGDL ATEYLQLMVY VTKTSIKIPS SHTLMKGGST KYWSGNSEFH FYSINVGGFF KLRSGEEISI EVSNPSLLDP DQDATYFGAF KVRDID.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14788
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 10mM sodium phosphate with no preservative; pH 7.5
Gene ID (Entrez) 8600
Molecular Weight (g/mol) ∽20
Name Human CD254 (RANK Ligand) (soluble)
pH Range 7.5
Quantity 10 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias H-RANK-L
Common Name Human CD254 (RANK Ligand) (soluble)
Gene Symbol TNFSF11
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitro research only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.