Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TRANCE (RANKL) (soluble) Recombinant Protein, PeproTech®

Catalog No. 31001C100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31001C100UG 100 μg
31001C10UG 10 μg
310-01C-1MG 1 mg
31001C250UG 250 μg
310-01C-2UG 2 μg
31001C500UG 500 μg
31001C50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 31001C100UG Supplier Gibco™ Supplier No. 31001C100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human sRANK Ligand is a 22.1 kDa polypeptide comprised of the TNF-homologous region of RANKL and a N-terminal His-tag. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene encodes a member of the tumor necrosis factor (TNF) cytokine family which is a ligand for osteoprotegerin and functions as a key factor for osteoclast differentiation and activation. This protein was shown to be a dentritic cell survival factor and is involved in the regulation of T cell-dependent immune response. T cell activation was reported to induce expression of this gene and lead to an increase of osteoclastogenesis and bone loss. This protein was shown to activate antiapoptotic kinase AKT/PKB through a signaling complex involving SRC kinase and tumor necrosis factor receptor-associated factor (TRAF) 6, which indicated this protein may have a role in the regulation of cell apoptosis. Targeted disruption of the related gene in mice led to severe osteopetrosis and a lack of osteoclasts. The deficient mice exhibited defects in early differentiation of T and B lymphocytes, and failed to form lobulo-alveolar mammary structures during pregnancy. Two alternatively spliced transcript variants have been found.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14788
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 8600
Molecular Weight (g/mol) 22.1 kDa
Name Human TRANCE (RANKL) (soluble)
Quantity 100 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CD254; H-RANK-L; hRANKL2; Ly109l; M-Rank-L; ODF; OPG; OPG ligand; OPGL; OPTB2; Osteoclast differentiation factor; osteoprotegerin ligand; Rankl; rank-l; receptor activator of NF-kappaB ligand; receptor activator of nuclear factor kappa B ligand; receptor activator of nuclear factor kappa-B ligand; RP11-86N24.2; sOdf; srankl; TNF-related activation-induced cytokine; Tnfsf11; TNLG6B; Trance; tumor necrosis factor (ligand) superfamily member 11; tumor necrosis factor (ligand) superfamily, member 11; tumor necrosis factor ligand 6 B; tumor necrosis factor ligand superfamily member 11; Tumor necrosis factor ligand superfamily member 11, membrane form; Tumor necrosis factor ligand superfamily member 11, soluble form; tumor necrosis factor superfamily member 11; tumor necrosis factor-related activation-induced cytokine
Common Name RANKL (RANK Ligand)
Gene Symbol TNFSF11
Biological Activity Determined by its dose-dependent ability to induce reporter gene in HT-29 NF-kappaB Luc reporter cells.
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence HHHHHHHHPS PGGSGGSQHI RAEKAMVDGS WLDLAKRSKL EAQPFAHLTI NATDIPSGSH KVSLSSWYHD RGWGKISNMT FSNGKLIVNQ DGFYYLYANI CFRHHETSGD LATEYLQLMV YVTKTSIKIP SSHTLMKGGS TKYWSGNSEF HFYSINVGGF FKLRSGEEIS IEVSNPSLLD PDQDATYFGA FKVRDID
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.