Learn More
Biomatik Corporation Human Transforming Growth Factor-Beta-Induced Protein Ig-H3 (Tgfbi/Bigh3) Elisa Kit
Supplier:  Biomatik Corporation EKC31342
Encompass_Preferred
Human Transforming Growth Factor-Beta-Induced Protein Ig-H3 (Tgfbi/Bigh3) Elisa Kit for detecting Transforming Growth Factor, Beta-Induced, 68Kda in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q15582; TGFBI. Target Synonyms: RGD containing collagen associated protein; AI181842; AI747162; Beta ig; Beta ig h3; Beta ig-h3; BGH3_HUMAN; Big h3; BIGH3; CDB1; CDG2; CDGG1; CSD; CSD1; CSD2; CSD3; EBMD; Kerato epithelin; Kerato-epithelin; LCD1; MGC150270; RGD CAP; RGD-CAP; RGD-containing collagen-associated protein; TGFBI; TGFBI transforming growth factor; beta induced; 68kDa; Transforming growth factor beta induced protein ig h3; Transforming growth factor-beta-induced protein ig-h3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9191
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.