Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TREM-1 Fc Recombinant Protein, PeproTech®

Catalog No. 31036250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31036250UG 250 μg
31036100UG 100 μg
310-36-10UG 10 μg
310-36-1MG 1 mg
31036500UG 500 μg
310-36-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 31036250UG Supplier Gibco™ Supplier No. 31036250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human TREM-1 Fc is a glycosylated, disulfide-linked homodimer of 826 amino acid residues whose monomer consists of the 180-amino-acid length extracellular portion of TREM-1 fused to the 231-amino-acid length Fc portion of human IgG1 by two glycines. The calculated molecular weight of Recombinant Human TREM-1 Fc dimer is 92.6 kDa; however, due to glycosylation, the monomer and dimer migrate at apparent molecular weights of approximately 60-65 kDa and 110-116 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TREM1 (triggering receptor expressed on myeloid cells 1) is a receptor belonging to the Ig superfamily that amplifies neutrophil and monocyte-mediated inflammatory responses triggered by bacterial and fungal infections by stimulating release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers. It is thought that TREM1 other related TREM proteins such as TREM2 act to modulate the inflammatory response. TREM1 has also been linked to inflammatory bowel disease resulting in enhanced production of TNF, IL-6, IL-8, and MCP-1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NP99
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 54210
Molecular Weight (g/mol) 92.6 kDa
Name Human TREM-1 Fc
Quantity 250 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CD354; sCD354; soluble CD354; soluble TREM 1; soluble TREM1; sTREM 1; sTREM1; TREM; Trem1; TREM-1; triggering receptor expressed in monocytes 1; triggering receptor expressed on monocytes 1; Triggering receptor expressed on myeloid cells; Triggering receptor expressed on myeloid cells 1
Common Name TREM1
Gene Symbol TREM1
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence ATKLTEEKYE LKEGQTLDVK CDYTLEKFAS SQKAWQIIRD GEMPKTLACT ERPSKNSHPV QVGRIILEDY HDHGLLRVRM VNLQVEDSGL YQCVIYQPPK EPHMLFDRIR LVVTKGFSGT PGSNENSTQN VYKIPPTTTK ALCPLYTSPR TVTQAPPKST ADVSTPDSEI NLTNVTDIIR GGPKSCDKTH TCPPCPAPEL LGGPSVFLFP PKPKDTLMIS RTPEVTCVVV DVSHEDPEVK FNWYVDGVEV HNAKTKPREE QYNSTYRVVS VLTVLHQDWL NGKEYKCKVS NKALPAPIEK TISKAKGQPR EPQVYTLPPS RDELTKNQVS LTCLVKGFYP SDIAVEWESN GQPENNYKTT PPVLDSDGSF FLYSKLTVDK SRWQQGNVFS CSVMHEALHN HYTQKSLSLS PGK
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.