Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIB3 Partial ORF (AAH27484, 56 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965629 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-629 25 μg
89-965-628 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-629 Supplier Abnova™ Supplier No. H00057761Q01L

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a putative protein kinase that is induced by the transcription factor NF-kappaB. The encoded protein is a negative regulator of NF-kappaB and can also sensitize cells to TNF- and TRAIL-induced apoptosis. In addition, this protein can negatively regulate the cell survival serine-threonine kinase AKT1. [provided by RefSeq]

Sequence: PDRATAVATASRLGPYVLLEPEEGGRAYRALHCPTGTEYTCKVYPVQEALAVLEPYARLPPHKHVARPTEVLAGTQLLYAFFTRTHGDMH

Specifications

Accession Number AAH27484
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57761
Molecular Weight (g/mol) 35.53kDa
Name TRIB3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen PDRATAVATASRLGPYVLLEPEEGGRAYRALHCPTGTEYTCKVYPVQEALAVLEPYARLPPHKHVARPTEVLAGTQLLYAFFTRTHGDMH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias C20orf97/NIPK/SINK/SKIP3/TRB3
Common Name TRIB3
Gene Symbol TRIB3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.