Learn More
Abnova™ Human TRIM10 Full-length ORF (AAH93926.1, 1 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic bodies. Studies in mice suggest that this protein plays a role in terminal differentiation of erythroid cells. Alternate splicing of this gene generates two transcript variants encoding different isoforms. [provided by RefSeq]
Specifications
Specifications
| Accession Number | AAH93926.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 10107 |
| Molecular Weight (g/mol) | 71.7kDa |
| Name | TRIM10 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | MASAASVTSLADEVNCPICQGTLREPVTIDCGHNFCRACLTRYCEIPGPDLEESPTCPLCKEPFRPGSFRPNWQLANVVENIERLQLVSTLGLGEEDVCQEHGEKIYFFCEDDEMQLCVVCREAGEHATHTMRFLEDAAAPYREQIHKCLKCLRKEREEIQEIQSRENKRMQVLLTQVSTKRQQVISEFAHLRKFLEEQQSILLAQLESQDGDILRQRDEFDLLVAGEICRFSALIEELEEKNERPARELLTDIRSTLIRCETRKCRKPVAVSPELGQRIRDFPQQALPLQREMKMFLEKLCFELDYEPAHISLDPQTSHPKLLLSEDHQRAQFSYKWQNSPDNPQRFDRATCVLAHTGITGGRHTWVWMARVPGDSGCCQFCSPPSVLGTEVAA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.