Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIM10 Partial ORF (NP_006769, 3 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965634 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-634 25 μg
89-965-633 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-634 Supplier Abnova™ Supplier No. H00010107Q01L

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic bodies. Studies in mice suggest that this protein plays a role in terminal differentiation of erythroid cells. Alternate splicing of this gene generates two transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: SAASVTSLADEVNCPICQGTLREPVTIDCGHNFCRACLTRYCEIPGPDLEESPTCPLCKEPFRPGSFRPNWQLANVVENIERLQLVSTLGLGEEDVCQEHGGKIYFFC

Specifications

Accession Number NP_006769
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10107
Molecular Weight (g/mol) 37.62kDa
Name TRIM10 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen SAASVTSLADEVNCPICQGTLREPVTIDCGHNFCRACLTRYCEIPGPDLEESPTCPLCKEPFRPGSFRPNWQLANVVENIERLQLVSTLGLGEEDVCQEHGGKIYFFC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HERF1/MGC141979/RFB30/RNF9
Common Name TRIM10
Gene Symbol TRIM10
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.