Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIM16 Partial ORF (NP_006461, 165 a.a. - 273 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965646 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-646 25 μg
89-965-645 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-646 Supplier Abnova™ Supplier No. H00010626Q01L

Used for AP, Array, ELISA, WB-Re

This gene was identified as an estrogen and anti-estrogen regulated gene in epithelial cells stably expressing estrogen receptor. The protein encoded by this gene contains two B box domains and a coiled-coiled region that are characteristic of the B box zinc finger protein family. The proteins of this family have been reported to be involved in a variety of biological processes including cell growth, differentiation and pathogenesis. Expression of this gene was detected in most tissues. Its function, however, has not yet been determined. [provided by RefSeq]

Sequence: LDAARRDKEAELQCTQLDLERKLKLNENAISRLQANQKSVLVSVSEVKAVAEMQFGELLAAVRKAQANVMLFLEEKEQAALSQANGIKAHLEYRSAEMEKSKQELERMA

Specifications

Accession Number NP_006461
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10626
Molecular Weight (g/mol) 37.73kDa
Name TRIM16 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen LDAARRDKEAELQCTQLDLERKLKLNENAISRLQANQKSVLVSVSEVKAVAEMQFGELLAAVRKAQANVMLFLEEKEQAALSQANGIKAHLEYRSAEMEKSKQELERMA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias EBBP
Common Name TRIM16
Gene Symbol TRIM16
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.