Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIM22 Full-length ORF (NP_006065.2, 1 a.a. - 498 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965659 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-659 25 μg
89-965-658 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-659 Supplier Abnova™ Supplier No. H00010346P01L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the cytoplasm and its expression is induced by interferon. The protein down-regulates transcription from the HIV-1 LTR promoter region, suggesting that function of this protein may be to mediate interferon's antiviral effects. [provided by RefSeq]

Sequence: MDFSVKVDIEKEVTCPICLELLTEPLSLDCGHSFCQACITAKIKESVIISRGESSCPVCQTRFQPGNLRPNRHLANIVERVKEVKMSPQEGQKRDVCEHHGKKLQIFCKEDGKVICWVCELSQEHQGHQTFRINEVVKECQEKLQVALQRLIKEDQEAEKLEDDIRQERTAWKNYIQIERQKILKGFNEMRVILDNEEQRELQKLEEGEVNVLDNLAAATDQLVQQRQDASTLISDLQRRLRGSSVEMLQDVIDVMKRSESWTLKKPKSVSKKLKSVFRVPDLSGMLQVLKELTDVQYYWVDVMLNPGSATSNVAISVDQRQVKTVRTCTFKNSNPCDFSAFGVFGCQYFSSGKYYWEVDVSGKIAWILGVHSKISSLNKRKSSGFAFDPSVNYSKVYSRYRPQYGYWVIGLQNTCEYNAFEDSSSSDPKVLTLFMAVPPCRIGVFLDYEAGIVSFFNVTNHGALIYKFSGCRFSRPAYPYFNPWNCLVPMTVCPPSS

Specifications

Accession Number NP_006065.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10346
Molecular Weight (g/mol) 83.3kDa
Name TRIM22 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MDFSVKVDIEKEVTCPICLELLTEPLSLDCGHSFCQACITAKIKESVIISRGESSCPVCQTRFQPGNLRPNRHLANIVERVKEVKMSPQEGQKRDVCEHHGKKLQIFCKEDGKVICWVCELSQEHQGHQTFRINEVVKECQEKLQVALQRLIKEDQEAEKLEDDIRQERTAWKNYIQIERQKILKGFNEMRVILDNEEQRELQKLEEGEVNVLDNLAAATDQLVQQRQDASTLISDLQRRLRGSSVEMLQDVIDVMKRSESWTLKKPKSVSKKLKSVFRVPDLSGMLQVLKELTDVQYYWVDVMLNPGSATSNVAISVDQRQVKTVRTCTFKNSNPCDFSAFGVFGCQYFSSGKYYWEVDVSGKIAWILGVHSKISSLNKRKSSGFAFDPSVNYSKVYSRYRPQYGYWVIGLQNTCEYNAFEDSSSSDPKVLTLFMAVPPCRIGVFLDYEAGIVSFFNVTNHGALIYKFSGCRFSRPAYPYFNPWNCLVPMTVCPPSS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GPSTAF50/RNF94/STAF50
Common Name TRIM22
Gene Symbol TRIM22
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.