Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIM25 Full-length ORF (AAH16924.1, 1 a.a. - 630 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965667 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-667 10 μg
89-965-668 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-667 Supplier Abnova™ Supplier No. H00007706P01S

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the cytoplasm. The presence of potential DNA-binding and dimerization-transactivation domains suggests that this protein may act as a transcription factor, similar to several other members of the TRIM family. Expression of the gene is upregulated in response to estrogen, and it is thought to mediate estrogen actions in breast cancer as a primary response gene. [provided by RefSeq]

Sequence: MAELCPLAEELSCSICLEPFKEPVTTPCGHNFCGSCLNETWAVQGSPYLCPQCRAVYQARPQLHKNTVLCNVVEQFLQADLAREPPADVWTPPARASAPSPNAQVACDHCLKEAAVKTCLVCMASFCQEHLQPHFDSPAFQDHPLQPPVRDLLRRKCSQHNRLREFFCPEHSECICHICLVEHKTCSPASLSQASADLEATLRHKLTVMYSQINGASRALDDVRNRQQDVRMTANRKVEQLQQEYTEMKALLDASETTSTRKIKEEEKRVNSKFDTIYQILLKKKSEIQTLKEEIEQSLTKRDEFEFLEKASKLRGISTKPVYIPEVELNHKLIKGIHQSTIDLKNELKQCIGRLQELTPSSGDPGEHDPASTHKSTRPVKKVSKEEKKSKKPPPVPALPSKLPTFGAPEQLVDLKQAGLEAAAKATSSHPNSTSLKAKVLETFLAKSRPELLEYYIKVILDYNTAHNKVALSECYTVASVAEMPQNYRPHPQRFTYCSQVLGLHCYKKGIHYWEVELQKNNFCGVGICYGSMNRQGPESRLGRNSASWCVEWFNTKISAWHNNVEKTLPSTKATRVGVLLNCDHGFVIFFAVADKVHLMYKFRVDFTEALYPAFWVFSAGATLSICSPK

Specifications

Accession Number AAH16924.1
For Use With (Application) Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7706
Molecular Weight (g/mol) 97.4kDa
Name Human TRIM25 Full-length ORF (AAH16924.1, 1 a.a. - 630 a.a.) Recombinant Protein with GST-tag at N-terminal
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Storage Requirements Store at −80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias EFP, RNF147, Z147, ZNF147
Common Name TRIM25
Gene Symbol TRIM25
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST Tag
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.