Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIM29 Partial ORF (NP_036233.2, 488 a.a. - 588 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965681 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-681 10 μg
89-965-682 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-681 Supplier Abnova™ Supplier No. H00023650Q01S

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene belongs to the TRIM protein family. It has multiple zinc finger motifs and a leucine zipper motif. It has been proposed to form homo- or heterodimers which are involved in nucleic acid binding. Thus, it may act as a transcriptional regulatory factor involved in carcinogenesis and/or differentiation. It may also function in the suppression of radiosensitivity since it is associated with ataxia telangiectasia phenotype. [provided by RefSeq]

Sequence: SSPGRFTKETTQKNFNNLYGTKGNYTSRVWEYSSSIQNSDNDLPVVQGSSSFSLKGYPSLMRSQSPKAQPQTWKSGKQTMLSHYRPFYVNKGNGIGSNEAP

Specifications

Accession Number NP_036233.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23650
Molecular Weight (g/mol) 36.85kDa
Name TRIM29 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen SSPGRFTKETTQKNFNNLYGTKGNYTSRVWEYSSSIQNSDNDLPVVQGSSSFSLKGYPSLMRSQSPKAQPQTWKSGKQTMLSHYRPFYVNKGNGIGSNEAP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ATDC/FLJ36085
Common Name TRIM29
Gene Symbol TRIM29
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.