Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIM32 Full-length ORF (AAH03154.1, 1 a.a. - 653 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965690 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-690 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-965-690 Supplier Abnova™ Supplier No. H00022954P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein has also been localized to the nucleus, where it interacts with the activation domain of the HIV-1 Tat protein. The Tat protein activates transcription of HIV-1 genes. [provided by RefSeq]

Sequence: MAAAAASHLNLDALREVLECPICMESFTEEQLRPKLLHCGHTICRQCLEKLLASSINGVRCPFCSKITRITSLTQLTDNLTVLKIIDTAGLSEAVGLLMCRSCGRRLPRQFCRSCGLVLCEPCREADHQPPGHCTLPVKEAAEERRRDFGEKLTRLRELMGELQRRKAALEGVSKDLQARYKAVLQEYGHEERRVQDELARSRKFFTGSLAEVEKSNSQVVEEQSYLLNIAEVQAVSRCDYFLAKIKQADVALLEETADEEEPELTASLPRELTLQDVELLKVGHVGPLQIGQAVKKPRTVNVEDSWAMEATASAASTSVTFREMDMSPEEVVASPRASPAKQRGPEAASNIQQCLFLKKMGAKGSTPGMFNLPVSLYVTSQGEVLVADRGNYRIQVFTRKGFLKEIRRSPSGIDSFVLSFLGADLPNLTPLSVAMNCQGLIGVTDSYDNSLKVYTLDGHCVACHRSQLSKPWGITALPSGQFVVTDVEGGKLWCFTVDRGSGVVKYSCLCSAVRPKFVTCDAEGTVYFTQGLGLNLENRQNEHHLEGGFSIGSVGPDGQLGRQISHFFSENEDFRCIAGMCVDARGDLIVADSSRKEILHFPKGGGYSVLIREGLTCPVGIALTPKGQLLVLDCWDHCIKIYSYHLRRYSTP

Specifications

Accession Number AAH03154.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 22954
Molecular Weight (g/mol) 98.4kDa
Name TRIM32 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MAAAAASHLNLDALREVLECPICMESFTEEQLRPKLLHCGHTICRQCLEKLLASSINGVRCPFCSKITRITSLTQLTDNLTVLKIIDTAGLSEAVGLLMCRSCGRRLPRQFCRSCGLVLCEPCREADHQPPGHCTLPVKEAAEERRRDFGEKLTRLRELMGELQRRKAALEGVSKDLQARYKAVLQEYGHEERRVQDELARSRKFFTGSLAEVEKSNSQVVEEQSYLLNIAEVQAVSRCDYFLAKIKQADVALLEETADEEEPELTASLPRELTLQDVELLKVGHVGPLQIGQAVKKPRTVNVEDSWAMEATASAASTSVTFREMDMSPEEVVASPRASPAKQRGPEAASNIQQCLFLKKMGAKGSTPGMFNLPVSLYVTSQGEVLVADRGNYRIQVFTRKGFLKEIRRSPSGIDSFVLSFLGADLPNLTPLSVAMNCQGLIGVTDSYDNSLKVYTLDGHCVACHRSQLSKPWGITALPSGQFVVTDVEGGKLWCFTVDRGSGVVKYSCLCSAVRPKFVTCDAEGTVYFTQGLGLNLENRQNEHHLEGGFSIGSVGPDGQLGRQISHFFSENEDFRCIAGMCVDARGDLIVADSSRKEILHFPKGGGYSVLIREGLTCPVGIALTPKGQLLVLDCWDHCIKIYSYHLRRYSTP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BBS11/HT2A/LGMD2H/TATIP
Common Name TRIM32
Gene Symbol TRIM32
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.