Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIM37 Partial ORF (NP_056109, 865 a.a. - 964 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965706 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-706 10 ug
89-965-707 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-706 Supplier Abnova™ Supplier No. H00004591Q01S

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the tripartite motif (TRIM) family, whose members are involved in diverse cellular functions such as developmental patterning and oncogenesis. The TRIM motif includes zinc-binding domains, a RING finger region, a B-box motif and a coiled-coil domain. The RING finger and B-box domains chelate zinc and might be involved in protein-protein and/or protein-nucleic acid interactions. The gene mutations are associated with mulibrey (muscle-liver-brain-eye) nanism, an autosomal recessive disorder that involves several tissues of mesodermal origin. Alternatively spliced transcript variants encoding the same protein have been identified. [provided by RefSeq]

Sequence: GHLEGLQMTDLENNSETGELQPVLPEGASAAPEEGMSSDSDIECDTENEEQEEHTSVGGFHDSFMVMTQPPDEDTHSSFPDGEQIGPEDLSFNTDENSGR

Specifications

Accession Number NP_056109
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4591
Molecular Weight (g/mol) 36.74kDa
Name TRIM37 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen GHLEGLQMTDLENNSETGELQPVLPEGASAAPEEGMSSDSDIECDTENEEQEEHTSVGGFHDSFMVMTQPPDEDTHSSFPDGEQIGPEDLSFNTDENSGR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KIAA0898/MUL/POB1/TEF3
Common Name TRIM37
Gene Symbol TRIM37
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.