Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIM61 Partial ORF (NP_001012414, 110 a.a. - 208 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965758 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-758 10 μg
89-965-759 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-758 Supplier Abnova™ Supplier No. H00391712Q01S

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for AP, Array, ELISA, WB-Re

Sequence: KDLELLCPRCSLSTDHQHHCVWPIKKAASYHRKKLEEYNAPWKERVELIEKVITMQTRKSLELKKKMESPSVTRLECSCTISAHFNLRLPGSSDSSASG

Specifications

Accession Number NP_001012414
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 391712
Molecular Weight (g/mol) 36.63kDa
Name TRIM61 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen KDLELLCPRCSLSTDHQHHCVWPIKKAASYHRKKLEEYNAPWKERVELIEKVITMQTRKSLELKKKMESPSVTRLECSCTISAHFNLRLPGSSDSSASG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC102996/RNF35
Common Name TRIM61
Gene Symbol TRIM61
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.