Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Tripeptidyl-peptidase I/TPP1 ELISA Kit

Catalog No. EH465RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH465RB 96 Tests
EH465RBX5 5 x 96 Tests
EH465RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH465RB Supplier Invitrogen™ Supplier No. EH465RB
Add to Cart
Add to Cart

ELISA

Human Tripeptidyl-peptidase I/TPP1 quantitates human Tripeptidyl-peptidase I/TPP1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Tripeptidyl-peptidase I/TPP1.

This gene encodes a member of the sedolisin family of serine proteases. The protease functions in the lysosome to cleave N-terminal tripeptides from substrates, and has weaker endopeptidase activity. It is synthesized as a catalytically-inactive enzyme which is activated and auto-proteolyzed upon acidification. Mutations in this gene result in late-infantile neuronal ceroid lipofuscinosis, which is associated with the failure to degrade specific neuropeptides and a subunit of ATP synthase in the lysosome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Cell growth-inhibiting gene 1 protein; ceroid-lipofuscinosis, neuronal 2; CLN 2; Cln2; GIG 1; GIG1; growth-inhibiting protein 1; LPIC; lysosomal pepstatin insensitive protease; Lysosomal pepstatin-insensitive protease; MGC21297; SCAR7; TPP 1; TPP I; TPP1; TPP-1; TPPI; TPP-I; tripeptidyl aminopeptidase; tripeptidyl peptidase 1; tripeptidyl peptidase I; tripeptidyl peptidase-I; tripeptidyl-peptidase 1; Tripeptidyl-peptidase I; UNQ267/PRO304
Assay Range 10 to 2,500 pg/mL
Assay Sensitivity 10 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 1200
Gene Symbol TPP1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CLN2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.