Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TrkA ELISA Kit

Catalog No. EH466RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH466RB 96 Tests
EH466RBX5 5 x 96 Tests
EH466RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH466RB Supplier Invitrogen™ Supplier No. EH466RB
Add to Cart
Add to Cart

ELISA

Human TrkA quantitates human TrkA in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human TrkA.

NTRK1 (TRKA) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. TrkA is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of TrkA leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number C80751; DKFZp781I14186; EC 2.7.10.1; gp140trk; high affinity nerve growth factor receptor; kinase TrkA; MTC; neurotrophic receptor tyrosine kinase 1; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; Ntrk1; Oncogene TRK; p140-TrkA; slow nerve growth factor receptor; Tkr; trk; TRK1; TRK1-transforming tyrosine kinase protein; TRKA; trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A
Assay Range 2 to 500 ng/mL
Assay Sensitivity 2 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4914
Gene Symbol Ntrk1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target TrkA
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.