Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Trypsin Pan/PRSS1-2-3 ELISA Kit

Catalog No. EH470RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH470RB 96 Tests
EH470RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH470RB Supplier Invitrogen™ Supplier No. EH470RB
Add to Cart
Add to Cart

ELISA

Human Trypsin Pan/PRSS1-2-3 quantitates human Trypsin Pan/PRSS1-2-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Trypsin Pan/PRSS1-2-3.

Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screene for and diagnose Cystic Fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Alpha-trypsin chain 1; Alpha-trypsin chain 2; Anionic trypsinogen; beta-trypsin; Brain trypsinogen; cationic trypsinogen; digestive zymogen; mesotrypsin; Mesotrypsinogen; MTG; nonfunctional trypsin 1; pancreatic trypsinogen III; protease serine 2 preproprotein; protease, serine 1; protease, serine 2; protease, serine 3; protease, serine, 2 (trypsin 2); protease, serine, 4 (trypsin 4, brain); PRSS1; PRSS2; PRSS3; PRSS4; Serine protease 1; Serine protease 2; Serine protease 3; Serine protease 4; T9; TCR V beta 4.1; TRP1; TRY1; TRY2; TRY3; TRY4; TRY8; TRYP1; TRYP2; Trypsin I; Trypsin II; trypsin III; Trypsin IV; trypsin-1; Trypsin-2; Trypsin-3; trypsinogen 1; trypsinogen 2; trypsinogen 4; trypsinogen 5; trypsinogen A; trypsinogen IV
Assay Range 30.72 to 7,500 pg/mL
Assay Sensitivity 30 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5644, 5645, 5646
Gene Symbol PRSS1, PRSS2, PRSS3
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Trypsin Pan
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.