Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TSC22D3 Full-length ORF (NP_932174.1, 1 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965908 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-908 10 ug
89-965-909 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-908 Supplier Abnova™ Supplier No. H00001831P02S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene shares significant sequence identity with the murine TSC-22 and Drosophila shs, both of which are leucine zipper proteins, that function as transcriptional regulators. The expression of this gene is stimulated by glucocorticoids and interleukin 10, and it appears to play a key role in the anti-inflammatory and immunosuppressive effects of this steroid and chemokine. Transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MAQSKLDCRSPVGLDCCNCCLDLAHRSGLQRGSSGENNNPGSPTVSNFRQLQEKLVFENLNTDKLNSIMRQDSLEPVLRDPCYLINEGICNRNIDQTMLSILLFFHSASGASVVAIDNKIEQAMDLVKNHLMYAVREEVEILKEQIRELVEKNSQLERENTLLKTLASPEQLEKFQSCLSPEEPAPESPQVPEAPGGSAV

Specifications

Accession Number NP_932174.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1831
Molecular Weight (g/mol) 48.6kDa
Name TSC22D3 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MAQSKLDCRSPVGLDCCNCCLDLAHRSGLQRGSSGENNNPGSPTVSNFRQLQEKLVFENLNTDKLNSIMRQDSLEPVLRDPCYLINEGICNRNIDQTMLSILLFFHSASGASVVAIDNKIEQAMDLVKNHLMYAVREEVEILKEQIRELVEKNSQLERENTLLKTLASPEQLEKFQSCLSPEEPAPESPQVPEAPGGSAV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DIP/DKFZp313A1123/DSIPI/GILZ/TSC-22R/hDIP
Common Name TSC22D3
Gene Symbol TSC22D3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.