Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TSEN2 Full-length ORF (NP_079541.1, 1 a.a. - 465 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965917 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-917 25 μg
89-965-916 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-917 Supplier Abnova™ Supplier No. H00080746P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes one of the subunits of the tRNA splicing endonuclease. This endonuclease catalyzes the first step in RNA splicing which is the removal of introns. Mutations in this gene have been associated with pontocerebellar hypoplasia type 2. A pseudogene has been identified on chromosome 4. Multiple transcript variants encoding different isoforms have been found for this gene

Sequence: MAEAVFHAPKRKRRVYETYESPLPIPFGQDHGPLKEFKIFRAEMINNNVIVRNAEDIEQLYGKGYFGKGILSRSRPSFTISDPKLVAKWKDMKTNMPIITSKRYQHSVEWAAELMRRQGQDESTVRRILKDYTKPLEHPPVKRNEEAQVHDKLNSGMVSNMEGTAGGERPSVVNGDSGKSGGVGDPREPLGCLQEGSGCHPTTESFEKSVREDASPLPHVCCCKQDALILQRGLHHEDGSQHIGLLHPGDRGPDHEYVLVEEAECAMSEREAAPNEELVQRNRLICRRNPYRIFEYLQLSLEEAFFLVYALGCLSIYYEKEPLTIVKLWKAFTVVQPTFRTTYMAYHYFRSKGWVPKVGLKYGTDLLLYRKGPPFYHASYSVIIELVDDHFEGSLRRPLSWKSLAALSRVSVNVSKELMLCYLIKPSTMTDKEMESPECMKRIKVQEVILSRWVSSRERSDQDDL

Specifications

Accession Number NP_079541.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 80746
Molecular Weight (g/mol) 79.6kDa
Name TSEN2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MAEAVFHAPKRKRRVYETYESPLPIPFGQDHGPLKEFKIFRAEMINNNVIVRNAEDIEQLYGKGYFGKGILSRSRPSFTISDPKLVAKWKDMKTNMPIITSKRYQHSVEWAAELMRRQGQDESTVRRILKDYTKPLEHPPVKRNEEAQVHDKLNSGMVSNMEGTAGGERPSVVNGDSGKSGGVGDPREPLGCLQEGSGCHPTTESFEKSVREDASPLPHVCCCKQDALILQRGLHHEDGSQHIGLLHPGDRGPDHEYVLVEEAECAMSEREAAPNEELVQRNRLICRRNPYRIFEYLQLSLEEAFFLVYALGCLSIYYEKEPLTIVKLWKAFTVVQPTFRTTYMAYHYFRSKGWVPKVGLKYGTDLLLYRKGPPFYHASYSVIIELVDDHFEGSLRRPLSWKSLAALSRVSVNVSKELMLCYLIKPSTMTDKEMESPECMKRIKVQEVILSRWVSSRERSDQDDL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC2776/MGC4440/PCH2B/SEN2/SEN2L
Common Name TSEN2
Gene Symbol TSEN2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.