Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TSFM Partial ORF (NP_005717, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965926 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-926 25 μg
89-965-925 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-926 Supplier Abnova™ Supplier No. H00010102Q01L

Used for AP, Array, ELISA, WB-Re

Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes. The TSFM gene encodes a mitochondrial translation elongation factor (Smeitink et al., 2006 [PubMed 17033963]).[supplied by OMIM]

Sequence: MSLLRSLRVFLVARTGSYPAGSLLRQSPQPRHTFYAGPRLSASASSKELLMKLRRKTGYSFVNCKKALETCGGDLKQAEIWLHKEAQKEGWSKAAKLQGR

Specifications

Accession Number NP_005717
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10102
Molecular Weight (g/mol) 36.74kDa
Name TSFM (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MSLLRSLRVFLVARTGSYPAGSLLRQSPQPRHTFYAGPRLSASASSKELLMKLRRKTGYSFVNCKKALETCGGDLKQAEIWLHKEAQKEGWSKAAKLQGR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias COXPD3/EF-TS/EF-Tsmt
Common Name TSFM
Gene Symbol TSFM
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.