Learn More
Gibco™ Human TSG Recombinant Protein, PeproTech®
Description
Recombinant Human TSG is a 198 amino acid, 22.2 kDa protein containing the BMP/TGF-beta binding portion of the full length TSG protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | Q9GZX9 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 57045 |
| Molecular Weight (g/mol) | 22.2 kDa |
| Name | Human TSG |
| Quantity | 250 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.