Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TSG Recombinant Protein, PeproTech®

Catalog No. 1200950UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
120-09-50UG 50 μg
12009100UG 100 μg
120-09-10UG 10 μg
120-09-1MG 1 mg
12009250UG 250 μg
12009500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 120-09-50UG Supplier Gibco™ Supplier No. 1200950UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human TSG is a 198 amino acid, 22.2 kDa protein containing the BMP/TGF-beta binding portion of the full length TSG protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TWSG1 may be involved in dorsoventral axis formation. Seems to antagonize BMP signaling by forming ternary complexes with CHRD and BMPs, thereby preventing BMPs from binding to their receptors. In addition to the anti-BMP function, also has pro-BMP activity, partly mediated by cleavage and degradation of CHRD, which releases BMPs from ternary complexes. May be an important modulator of BMP-regulated cartilage development and chondrocyte differentiation. May play a role in thymocyte development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9GZX9
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 57045
Molecular Weight (g/mol) 22.2 kDa
Name Human TSG
Quantity 50 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 1810013J15Rik; 9030422N06Rik; AW552143; D17Ertd403e; PSEC0250; Tsg; Twg; twisted gastrulation BMP signaling modulator 1; twisted gastrulation homolog 1; twisted gastrulation homolog 1 (Drosophila); twisted gastrulation protein homolog 1; Twsg1
Common Name TSG
Gene Symbol Twsg1
Biological Activity Determined by its ability to neutralize BMP-6-induced alkaline phosphatase production by ATDC chondrogenic cells. The expected ED50 for this effect is ≤ 2.0 ug/ml of TSG.
Conjugate Unconjugated
Recombinant Recombinant
Sequence CNKALCASDV SKCLIQELCQ CRPGEGNCSC CKECMLCLGA LWDECCDCVG MCNPRNYSDT PPTSKSTVEE LHEPIPSLFR ALTEGDTQLN WNIVSFPVAE ELSHHENLVS FLETVNQPHH QNVSVPSNNV HAPYSSDKEH MCTVVYFDDC MSIHQCKISC ESMGASKYRW FHNACCECIG PECIDYGSKT VKCMNCMF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.