Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TSKS Full-length ORF (AAH58862.1, 1 a.a. - 589 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965950 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-950 25 μg
89-965-949 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-950 Supplier Abnova™ Supplier No. H00060385P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene may play a role in testicular physiology, spermatogenesis or spermiogenesis. Expression of the encoded protein is highest in the testis and down-regulated in testicular cancer. The gene is localized to the region 19q13.3 among the related RAS viral oncogene homolog (RRAS) and interferon regulatory factor 3 (IRF3) genes, which are both involved in tumorigenesis pathways and progression. [provided by RefSeq]

Sequence: MASVVVKTIWQSKEIHEAGDTPTGVESCSQLVPEAPRRVTSRAKGIPKKKKAVSFHGVEPQMSHQPMHWCLNLKRSSACTNVSLLNLAAMEPTDSTGTDSTVEDLSGQLTLAGPPASPTLPWDPDDADITEILSGVNSGLVRAKDSITSLKEKTNRVNQHVQSLQSECSVLSENLERRRQEAEELEGYCIQLKENCWKVTRSVEDAEIKTNVLKQNSALLEEKLRYLQQQLQDETPRRQEAELQEPEEKQEPEEKQEPEEKQKPEAGLSWNSLGPAATSQGCPGPPGSPDKPSRPHGLVPAGWGMGPRAGEGPYVSEQELQKLFTGIEELRREVSSLTARWHQEEGAVQEALRLLGGLGGRVDGFLGQWERAQREQAQTARDLQELRGRADELCTMVERSAVSVASLRSELEGLGPLKPILEEFGRQFQNSRRGPDLSMNLDRSHQGNCARCASQGSQLSTESLQQLLDRALTSLVDEVKQRGLTPACPSCQRLHKKILELERQALAKHVRAEALSSTLRLAQDEALRAKNLLLTDKMKPEEKMATLDHLHLKMCSLHDHLSNLPLEGSTGTMGGGSSAGTPPKQGGSA

Specifications

Accession Number AAH58862.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 60385
Molecular Weight (g/mol) 91.1kDa
Name TSKS (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MASVVVKTIWQSKEIHEAGDTPTGVESCSQLVPEAPRRVTSRAKGIPKKKKAVSFHGVEPQMSHQPMHWCLNLKRSSACTNVSLLNLAAMEPTDSTGTDSTVEDLSGQLTLAGPPASPTLPWDPDDADITEILSGVNSGLVRAKDSITSLKEKTNRVNQHVQSLQSECSVLSENLERRRQEAEELEGYCIQLKENCWKVTRSVEDAEIKTNVLKQNSALLEEKLRYLQQQLQDETPRRQEAELQEPEEKQEPEEKQEPEEKQKPEAGLSWNSLGPAATSQGCPGPPGSPDKPSRPHGLVPAGWGMGPRAGEGPYVSEQELQKLFTGIEELRREVSSLTARWHQEEGAVQEALRLLGGLGGRVDGFLGQWERAQREQAQTARDLQELRGRADELCTMVERSAVSVASLRSELEGLGPLKPILEEFGRQFQNSRRGPDLSMNLDRSHQGNCARCASQGSQLSTESLQQLLDRALTSLVDEVKQRGLTPACPSCQRLHKKILELERQALAKHVRAEALSSTLRLAQDEALRAKNLLLTDKMKPEEKMATLDHLHLKMCSLHDHLSNLPLEGSTGTMGGGSSAGTPPKQGGSA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias TSKS1
Common Name TSKS
Gene Symbol TSKS
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.